
Chlorpromazine impurity A

Synonym(s):
Chlorpromazine sulfoxide, 3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1- amine S-oxide
Empirical Formula (Hill Notation):
C17H19ClN2OS
CAS Number:
Molecular Weight:
334.86
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing