
Carbamazepine 10,11-epoxide

Synonym(s):
1a,10b-Dihydro-6H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide
Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing