8arm-PEG20K-Acrylate, hexaglycerol core
Synonym(s):
8arm-PEG-Acrylate, hexaglycerol core
Linear Formula:
R(O(CH2CH2O)nCH2CH2OOCCHCH2)8
Products
Brand
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing