
4-tert-Octylphenol-3,5-d2 solution

Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol-3,5-d2, 4-tert-OP-D2
Empirical Formula (Hill Notation):
C14D2H20O
Products
Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing