Skip to Content
Merck
3-(Maleimido)propionic acid N-hydroxysuccinimide ester ≥98.5% (HPLC)

3-Maleimidopropionic acid N-hydroxysuccinimide ester

3-(Maleimido)propionic acid N-hydroxysuccinimide ester ≥98.5% (HPLC)
Synonym(s):
N-(3-Maleimidopropionyloxy)succinimide, N-Maleoyl-β-alanine N′-hydroxysuccinimide ester, N-Maleoyl-β-alanine N-hydroxysuccinimide ester, N-Succinimidyl 3-maleimidopropionate, BMPS, 3-(Maleimido)propionic acid N-succinimidyl ester, N-(3-Maleimidopropionyloxy)succinimide, N-Maleoyl-β-alanine N′-hydroxysuccinimide ester, N-Succinimidyl 3-maleimidopropionate
Empirical Formula (Hill Notation):
C11H10N2O6
CAS Number:
Molecular Weight:
266.21

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Reagent Type