Skip to Content
Merck
1,2,3,4,5-Pentamethylcyclopentadiene 95%

1,2,3,4,5-Pentamethylcyclopentadiene

1,2,3,4,5-Pentamethylcyclopentadiene 95%
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing