Key Documents
HS120551
Magnetic Stir Bar
L × diam. 30 mm × 8 mm
About This Item
UNSPSC Code:
41122401
NACRES:
NB.72
Pricing and availability is not currently available.
material
(with Pivot ring)
octagonal
round white PTFE stir bar(s)
packaging
pack of 10 ea
manufacturer/tradename
Heathrow Scientific 120551
L × diam.
30 mm × 8 mm
General description
The ALNICO magnetic stir bar withstands temperatures up to 250°C, making them suitable for use with both unheated and heated stirrers. Low friction octagon shape with a center pivot ring helps create a strong mixing vortex, even with more viscous solutions
Features and Benefits
  • Magnetic bar is made from ALNICO, one of the strongest permanent magnets available
  • The sintered manufacturing process reduces the chance of decoupling (spin outs), which weaker stir bars often suffer from
  • Encapsulation in PTFE protects both magnet and what it is stirring
  • Chemically inert and easy to clean
  • Can withstand temperatures up to 250°C, making them suitable for use with both unheated and heated stirrers
  • Low friction octagon shape with a center pivot ring helps create a strong mixing vortex, even with more viscous solutions
Choose from one of the most recent versions:
Certificates of Analysis (COA)