HPLC Analysis of Glutathione S-Transferase on TSKgel® G3000SWXL
![HPLC Analysis of Glutathione S-Transferase on TSKgel® G3000SWXL application for HPLC](/deepweb/assets/sigmaaldrich/product/images/206/257/d428943c-1560-4db9-aed0-2b6ba11ea35e/800/d428943c-1560-4db9-aed0-2b6ba11ea35e.jpg)
Materials
Application
Product No.
Description
Pricing
CONDITIONS
column
TSKgel G3000SWXL, 30 cm × 7.8 mm I.D., 5 μm particles (808541)
mobile phase
0.3M sodium chloride/0.05M sodium dihydrogen phosphate, pH 7.0
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220nm and enzyme assay
injection
100μL crude glutathione S-transferase preparation from guinea pig liver (0.7mg)
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation