Merck
  • 2H-Dinaphthopentacene: A Polycyclic Aromatic Hydrocarbon Core for Metal-Free Organic Sensitizers in Efficient Dye-Sensitized Solar Cells.
2H-Dinaphthopentacene: A Polycyclic Aromatic Hydrocarbon Core for Metal-Free Organic Sensitizers in Efficient Dye-Sensitized Solar Cells.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2017-09-22)
Yameng Ren, Jiao Liu, Aibin Zheng, Xiandui Dong, Peng Wang
ABSTRACT
Continuous studies on the use of a polycyclic aromatic hydrocarbon as the central block of an organic photosensitizer have brought forth a new opportunity toward efficiency enhancement of dye-sensitized solar cells (DSCs). In this paper, a nonacyclic aromatic hydrocarbon 9,19-dihydrodinaphtho[3,2,1-de:3',2',1'-op]pentacene, tethered with four 4-hexylphenyl solubilizing groups is reported. The novel chromophore 9,9-19-19-tetrakis(4-hexylphenyl)-9,19-dihydrodinaphtho[3,2,1-de:3',2',1'-op]pentacene is further functionalized with diarylamines and 4-(7-ethynylbenzo[c][1,2,5]thiadiazol-4-yl)benzoic acid to produce two donor-acceptor (D-A) organic photosensitizers, achieving good power conversion efficiencies up to 10.2% in DSCs.
Sigma-Aldrich
(Triisopropylsilyl)acetylene, 97%
Sigma-Aldrich
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II)