Merck
  • Lanthanide doped ultrafine hybrid nanostructures: multicolour luminescence, upconversion based energy transfer and luminescent solar collector applications.
Lanthanide doped ultrafine hybrid nanostructures: multicolour luminescence, upconversion based energy transfer and luminescent solar collector applications.
Nanoscale (2016-12-14)
Priyam Singh, Praveen Kumar Shahi, Sunil Kumar Singh, Akhilesh Kumar Singh, Manish Kumar Singh, Rajiv Prakash, Shyam Bahadur Rai
ABSTRACT
We herein demonstrate novel inorganic-organic hybrid nanoparticles (HNPs) composed of inorganic NPs, NaY
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Europium(III) oxide, 99.999% trace metals basis