Merck
CacyBP/SIP as a novel modulator of the thin filament.
Biochimica et biophysica acta (2012-12-26)
Ewelina Jurewicz, Zofia Ostrowska, Jolanta Jozwiak, Maria Jolanta Redowicz, Wieslawa Lesniak, Joanna Moraczewska, Anna Filipek
ABSTRACT
The CacyBP/SIP protein interacts with several targets, including actin. Since the majority of actin filaments are associated with tropomyosin, in this work we characterized binding of CacyBP/SIP to the actin-tropomyosin complex and examined the effects of CacyBP/SIP on actin filament functions. By using reconstituted filaments composed of actin and AEDANS-labeled tropomyosin, we observed that binding of CacyBP/SIP caused an increase in tropomyosin fluorescence intensity indicating the occurrence of conformational changes within the filament. We also found that CacyBP/SIP bound directly to tropomyosin and that these proteins did not compete with each other for binding to actin. Electron microscopy showed that in the absence of tropomyosin CacyBP/SIP destabilized actin filaments, but tropomyosin reversed this effect. Actin-activated myosin S1 ATPase activity assays, performed using a colorimetric method, indicated that CacyBP/SIP reduced ATPase activity and that the presence of tropomyosin enhanced this inhibitory effect. Thus, our results suggest that CacyBP/SIP, through its interaction with both actin and tropomyosin, regulates the organization and functional properties of the thin filament.
Sigma-Aldrich
Monoclonal Anti-Tropomyosin antibody produced in mouse, clone TM311, ascites fluid