Merck
  • Tumor suppressor SCUBE2 inhibits breast-cancer cell migration and invasion through the reversal of epithelial-mesenchymal transition.
Tumor suppressor SCUBE2 inhibits breast-cancer cell migration and invasion through the reversal of epithelial-mesenchymal transition.
Journal of cell science (2013-11-12)
Yuh-Charn Lin, Yi-Ching Lee, Ling-Hui Li, Chien-Jui Cheng, Ruey-Bing Yang
ABSTRACT
Signal peptide-CUB-EGF domain-containing protein 2 (SCUBE2) belongs to a secreted and membrane-associated multi-domain SCUBE protein family. We previously demonstrated that SCUBE2 is a novel breast-tumor suppressor and could be a useful prognostic marker. However, the role of SCUBE2 in breast-cancer cell migration and invasion and how it is regulated during the epithelial-mesenchymal transition (EMT) remain undefined. In this study, we showed that ectopic SCUBE2 overexpression could enhance the formation of E-cadherin-containing adherens junctions by β-catenin-SOX-mediated induction of forkhead box A1 (a positive regulator of E-cadherin) and upregulation of E-cadherin, which in turn led to epithelial transition and inhibited migration and invasion of aggressive MDA-MB-231 breast-carcinoma cells. SCUBE2 expression was repressed together with that of E-cadherin in TGF-β-induced EMT; direct expression of SCUBE2 alone was sufficient to inhibit the TGF-β-induced EMT. Furthermore, quantitative DNA methylation, methylation-specific PCR, and chromatin immunoprecipitation analyses revealed that SCUBE2 expression was inactivated by DNA hypermethylation at the CpG islands by recruiting and binding DNA methyltransferase 1 during TGF-β-induced EMT. Together, our results suggest that SCUBE2 plays a key role in suppressing breast-carcinoma-cell mobility and invasiveness by increasing the formation of the epithelial E-cadherin-containing adherens junctions to promote epithelial differentiation and drive the reversal of EMT.
Sigma-Aldrich
5-Aza-2′-Deoxycytidine, A cytosine analog that acts as a DNA methyltransferase inhibitor.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Trichostatin A, Streptomyces sp., A potent and reversible, cell-permeable inhibitor of histone deacetylase.