Merck
  • Release of tetracycline hydrochloride from electrospun poly(ethylene-co-vinylacetate), poly(lactic acid), and a blend.
Release of tetracycline hydrochloride from electrospun poly(ethylene-co-vinylacetate), poly(lactic acid), and a blend.
Journal of controlled release : official journal of the Controlled Release Society (2002-05-07)
El-Refaie Kenawy, Gary L Bowlin, Kevin Mansfield, John Layman, David G Simpson, Elliot H Sanders, Gary E Wnek
ABSTRACT
Electrospun fiber mats are explored as drug delivery vehicles using tetracycline hydrochloride as a model drug. The mats were made either from poly(lactic acid) (PLA), poly(ethylene-co-vinyl acetate) (PEVA), or from a 50:50 blend of the two. The fibers were electrospun from chloroform solutions containing a small amount of methanol to solubilize the drug. The release of the tetracycline hydrochloride from these new drug delivery systems was followed by UV-VIS spectroscopy. Release profiles from the electrospun mats were compared to a commercially available drug delivery system, Actisite (Alza Corporation, Palo Alto, CA), as well as to cast films of the various formulations.
Sigma-Aldrich
Trizma® hydrochloride solution, 1 M, BioReagent