Merck
  • Risk factors for obstetric morbidity in patients with uterine atony undergoing caesarean delivery.
Risk factors for obstetric morbidity in patients with uterine atony undergoing caesarean delivery.
British journal of anaesthesia (2014-06-08)
A J Butwick, B Carvalho, Y Y El-Sayed
ABSTRACT
Uterine atony (UA) is recognized as a leading cause of postpartum haemorrhage. However, knowledge of risk factors of haemorrhage-related morbidity among patients diagnosed with UA is uncertain. We investigated risk factors for haemorrhage-related morbidity among patients undergoing Caesarean delivery with UA. We conducted a secondary analysis of data sourced from a 4-yr observational study at 19 US academic centres. Patients with UA were identified based on receiving methylergonovine or carboprost. Our primary outcome (haemorrhage-related morbidity) included a composite of intra- or postpartum transfusion; Caesarean hysterectomy; uterine or hypogastric artery ligation; intensive care admission for: pulmonary oedema, coagulopathy, adult respiratory distress syndrome, postoperative ventilation, or invasive line monitoring. Among 57,182 patients who underwent Caesarean delivery, 2294 (4%) patients developed UA. Haemorrhage-related morbidity occurred in 450 (19.6%) patients with UA. The risk of haemorrhage-related morbidity was increased among African-Americans [adjusted odds ratio (aOR)=2.36; 95% confidence interval (CI)=1.73-3.23], Hispanics (aOR=1.4; 95% CI=1.04-1.9), women with multiple gestations (aOR=1.59; 95% CI=1.06-2.38), placenta praevia (aOR=4.89; 95% CI=3.04-7.87), patients with ASA class III (aOR=1.4; 95 CI=1.03-1.9), or ASA class IV (aOR=5.88; 95% CI=2.48-13.9), exposure to general anaesthesia (GA) (aOR=2.4; 95% CI=1.59-3.62) and combined general and regional anaesthesia (aOR=4.0; 95% CI=2.62-6.09), and ≥2 prior Caesarean deliveries (aOR=1.62; 95% CI=1.1-2.39). Among patients with UA undergoing Caesarean delivery, the risk of haemorrhage-related morbidity is increased in African-Americans, Hispanics, patients with multiple gestations, placenta praevia, ASA class III or IV, ≥2 prior Caesarean deliveries and those undergoing GA.
Product Description
USP
Methylergonovine maleate, United States Pharmacopeia (USP) Reference Standard
Methylergometrine for system suitability, European Pharmacopoeia (EP) Reference Standard
Methylergometrine maleate, European Pharmacopoeia (EP) Reference Standard