Merck
  • p38 kinase inhibitors for the treatment of arthritis and osteoporosis: thienyl, furyl, and pyrrolyl ureas.
p38 kinase inhibitors for the treatment of arthritis and osteoporosis: thienyl, furyl, and pyrrolyl ureas.
Bioorganic & medicinal chemistry letters (2001-01-05)
A M Redman, J S Johnson, R Dally, S Swartz, H Wild, H Paulsen, Y Caringal, D Gunn, J Renick, M Osterhout, J Kingery-Wood, R A Smith, W Lee, J Dumas, S M Wilhelm, T J Housley, A Bhargava, G E Ranges, A Shrikhande, D Young, M Bombara, W J Scott
ABSTRACT
Inhibitors of the MAP kinase p38 are potentially useful for the treatment for osteoporosis, arthritis, and other inflammatory diseases. A series of thienyl, furyl, and pyrrolyl ureas has been identified as potent p38 inhibitors, displaying in vitro activity in the nanomolar range.
Sigma-Aldrich
Methyl 3-amino-2-thiophenecarboxylate, 99%
Pricing and availability is not currently available.