Merck
  • Synthesis of a 2,4,6-trisubstituted 5-cyano-pyrimidine library and evaluation of its immunosuppressive activity in a Mixed Lymphocyte Reaction assay.
Synthesis of a 2,4,6-trisubstituted 5-cyano-pyrimidine library and evaluation of its immunosuppressive activity in a Mixed Lymphocyte Reaction assay.
Bioorganic & medicinal chemistry (2013-01-26)
Alessandro Stella, Kristien Van Belle, Steven De Jonghe, Thierry Louat, Jean Herman, Jef Rozenski, Mark Waer, Piet Herdewijn
ABSTRACT
A series of novel pyrimidine analogues were synthesized and evaluated for immunosuppressive activity in the Mixed Lymphocyte Reaction assay, which is well-known as the in vitro model for in vivo rejection after organ transplantation. Systematic variation of the substituents at positions 2, 4 and 6 of the pyrimidine scaffold led to the discovery of 2-benzylthio-5-cyano-6-(4-methoxyphenyl)-4-morpholinopyrimidine with an IC(50) value of 1.6 μM in the MLR assay.
Product Description
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Sigma-Aldrich
Pyrimidine, ≥98.0%
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm