Merck
  • Induction of glutathione S-transferase pi as a bioassay for the evaluation of potency of inhibitors of benzo(a)pyrene-induced cancer in a murine model.
Induction of glutathione S-transferase pi as a bioassay for the evaluation of potency of inhibitors of benzo(a)pyrene-induced cancer in a murine model.
International journal of cancer (1997-12-17)
X Hu, P J Benson, S K Srivastava, H Xia, R J Bleicher, H A Zaren, S Awasthi, Y C Awasthi, S V Singh
ABSTRACT
There is a growing need for short-term and cost-effective bioassay to assess the efficacy of potential chemo-preventive agents. We report that the induction of glutathione (GSH) S-transferase pi (mGSTP1-1) by a chemo-preventive agent can be used as a reliable marker to assess its efficacy in retarding chemical carcinogenesis induced by benzo(a)pyrene (BP), which is a widespread environmental pollutant and believed to be a risk factor in human chemical carcinogenesis. This conclusion is based on 1) the relative contribution of mGSTP1-1 of the liver and forestomach of female A/J mice in the detoxification of the ultimate carcinogenic metabolite of BP, (+)-anti-7,8-dihydroxy-9, 10-oxy-7,8,9, 10-tetrahydrobenzo(a)pyrene [(+)-anti-BPDE]; and 2) a positive correlation between the induction of hepatic and forestomach mGSTP1-1 by 5 naturally occurring organosulfides (OSCs) from garlic (diallyl sulfide, diallyl disulfide, diallyl trisulfide, dipropyl sulfide and dipropyl disulfide) and their effectiveness in preventing BP-induced forestomach neoplasia in mice. In the liver, the combined contribution of other GSTs in the detoxification of (+)-anti-BPDE was far less than the contribution of mGSTP1-1 alone. Likewise, in the forestomach, the contribution of mGSTP1-1 far exceeded the combined contribution of other GSTs. Studies on the effects of OSCs against BP-induced forestomach neoplasia revealed a good correlation between their chemo-preventive efficacy and their ability to induce mGSTP1-1 expression in the liver (r = -0.89; p < 0.05) as well as in the forestomach (r = -0.97; p < 0.05). Our results suggest that the induction of mGSTP1-1 may be a reliable marker for evaluating the efficacy of potential inhibitors of BP-induced cancer in a murine model.
Sigma-Aldrich
Propyl disulfide, ≥98%, FG
Sigma-Aldrich
Dipropyl disulfide, 98%
Sigma-Aldrich
Dipropyl sulfide, 97%