Merck
  • Decomposition of two types of electric wires considering the effect of the metal in the production of pollutants.
Decomposition of two types of electric wires considering the effect of the metal in the production of pollutants.
Chemosphere (2012-12-13)
Juan A Conesa, Silvia Egea, Julia Moltó, Nuria Ortuño, Rafael Font
ABSTRACT
Combustion runs at 700 °C in a horizontal laboratory furnace were carried out on two different electric wires (PVC and halogen-free wire). Tests were performed in the presence and in the absence of the metal conductor of the wires. The analyses of the polycyclic aromatic hydrocarbons (PAHs), chlorobenzenes (CBzs), chlorophenols (CPhs), mono- to octa-chlorodibenzo-p-dioxin and dibenzofurans (PCDD/Fs), and dioxin-like PCBs are shown. Regarding semivolatile compounds, PAHs production decreases in the presence of metal, while a higher amount of chlorinated compounds are emitted. Respect to the PCDD/Fs, the PVC wire in the presence of metal presents the highest emission, with a much more emission of furans than dioxins. The maximum emission is with 2 or 3 chlorine atom PCDD/Fs. PCBs emission correlates with PCDD/F production and represents 3-4% of total toxicity, determined by using WHO2005 factors.
Sigma-Aldrich
Dibenzofuran, 98%
Dibenzo[b,d]furan, BCR®, certified reference material