Merck
  • A severe case of Aeromonas veronii biovar sobria travellers' diarrhoea characterized by Vibrio parahaemolyticus co-isolation.
A severe case of Aeromonas veronii biovar sobria travellers' diarrhoea characterized by Vibrio parahaemolyticus co-isolation.
Journal of medical microbiology (2012-09-25)
Donatella Ottaviani, Francesca Leoni, Elena Rocchegiani, Sabrina Santarelli, Laura Masini, Maria Letizia D'Annibale, Anna Pianetti, Antonio Carraturo
ABSTRACT
We report a severe case of travellers' diarrhoea in a patient returning from Ecuador to Italy with the concomitant presence of Aeromonas veronii biovar sobria and Vibrio parahaemolyticus in their faeces. Based on diagnostic results, epidemiological information and the clinical outcome, we conclude that the real aetiological agent was A. veronii biovar sobria, while V. parahaemolyticus was only transient in the intestine of the patient.
Product Description
Acebutolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Acebutolol hydrochloride, analytical standard