Merck
  • Molecular recognition of saccharides by proteins. Insights on the origin of the carbohydrate-aromatic interactions.
Molecular recognition of saccharides by proteins. Insights on the origin of the carbohydrate-aromatic interactions.
Journal of the American Chemical Society (2005-05-19)
María del Carmen Fernández-Alonso, Francisco Javier Cañada, Jesús Jiménez-Barbero, Gabriel Cuevas
ABSTRACT
The existence of stabilizing carbohydrate-aromatic interactions is demonstrated from both the theoretical and experimental viewpoints. The geometry of experimentally based galactose-lectin complexes has been properly accounted for by using a MP2/6-31G(d,p) level of theory and by considering a counterpoise correction during optimization. In this case, the stabilizing interaction energy of the fucose-benzene complex amounts to 3.0 kcal/mol. The theoretical results obtained herein indicate that the carbohydrate-aromatic interactions are stabilizing interactions with an important dispersive component and that electronic density between the sugar hydrogens and the aromatic ring indeed exists, thus giving rise to three so-called nonconventional hydrogen bonds. Experimental evidence of the intrinsic tendency of aromatic moieties to interact with certain sugars has also been shown by simple NMR experiments in water solution. Benzene and phenol specifically interact with the clusters of C-H bonds of the alpha face of methyl beta-galactoside, without requiring the well-defined three-dimensional shape provided by a protein receptor, therefore resembling the molecular recognition features that are frequently observed in many carbohydrate-protein complexes.
Sigma-Aldrich
Methyl α-D-galactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Methyl-β-D-galactopyranoside