Merck
  • Treatment with Synthetic Pseudoceramide Improves Atopic Skin, Switching the Ceramide Profile to a Healthy Skin Phenotype.
Treatment with Synthetic Pseudoceramide Improves Atopic Skin, Switching the Ceramide Profile to a Healthy Skin Phenotype.
The Journal of investigative dermatology (2020-02-06)
Koichi Ishida, Akihiko Takahashi, Kotatsu Bito, Zoe Draelos, Genji Imokawa
ABSTRACT
Little is known about the pathophysiological linkages between altered ceramide profiles in the stratum corneum (SC) of patients with atopic dermatitis and their impaired skin barrier and water-holding functions. We studied those characteristics following topical treatment with a designed synthetic pseudoceramide (pCer) and analyzed that pathophysiological linkage by microanalyzing ceramides using normal phase liquid chromatography-electrospray ionization mass spectrometry. Four weeks of treatment with pCer significantly reduced skin symptoms, accompanied by significant decreases in transepidermal water loss and increases in water content. In the SC ceramide profiles, ceramides containing nonhydroxy fatty acids and 6-hydroxysphingosines (Cer[NH]) and ceramides containing nonhydroxy fatty acids and phytosphingosines (Cer[NP]) increased, whereas ceramides containing nonhydroxy fatty acids and sphingosines (Cer[NS]) and ceramides containing a-hydroxy fatty acids and sphingosines (Cer[AS]) decreased, with larger alkyl chain lengths in Cer[NS], distinctly representing a switch from an atopic dermatitis to a healthy skin phenotype. The level of pCer that penetrated into the SC was significantly correlated with the SC water content but not with transepidermal water loss. The level and the average carbon chain length of Cer[NS] were closely correlated with the pCer level in the SC. These findings indicate that the penetrated pCer contributes to shift the ceramide profile from an atopic dermatitis to a healthy skin phenotype. Taken together, the observed clinical efficacy of treatment with pCer provides a deep insight into the pathogenesis of atopic dermatitis as a ceramide-deficient disease.
Product Description
Avanti
C18 Ceramide (d17:1/18:0), Avanti Research - A Croda Brand 860646P, powder