Merck
  • The Origins and Vulnerabilities of Two Transmissible Cancers in Tasmanian Devils.
The Origins and Vulnerabilities of Two Transmissible Cancers in Tasmanian Devils.
Cancer cell (2018-04-11)
Maximilian R Stammnitz, Tim H H Coorens, Kevin C Gori, Dane Hayes, Beiyuan Fu, Jinhong Wang, Daniel E Martin-Herranz, Ludmil B Alexandrov, Adrian Baez-Ortega, Syd Barthorpe, Alexandra Beck, Francesca Giordano, Graeme W Knowles, Young Mi Kwon, George Hall, Stacey Price, Ruth J Pye, Jose M C Tubio, Hannah V T Siddle, Sukhwinder Singh Sohal, Gregory M Woods, Ultan McDermott, Fengtang Yang, Mathew J Garnett, Zemin Ning, Elizabeth P Murchison
ABSTRACT
Transmissible cancers are clonal lineages that spread through populations via contagious cancer cells. Although rare in nature, two facial tumor clones affect Tasmanian devils. Here we perform comparative genetic and functional characterization of these lineages. The two cancers have similar patterns of mutation and show no evidence of exposure to exogenous mutagens or viruses. Genes encoding PDGF receptors have copy number gains and are present on extrachromosomal double minutes. Drug screening indicates causative roles for receptor tyrosine kinases and sensitivity to inhibitors of DNA repair. Y chromosome loss from a male clone infecting a female host suggests immunoediting. These results imply that Tasmanian devils may have inherent susceptibility to transmissible cancers and present a suite of therapeutic compounds for use in conservation.
Sigma-Aldrich
Monoclonal Anti-YAP1 antibody produced in mouse, clone 2F12, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-TAZ antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PRX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution