Skip to Content
Merck
Tridodecylmethylammonium iodide 97%

Tridodecylmethylammonium iodide

Tridodecylmethylammonium iodide 97%
Synonym(s):
Methyltridodecylammonium iodide
Linear Formula:
[CH3(CH2)11]3N(I)CH3
CAS Number:
Molecular Weight:
663.93

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing