Skip to Content
Merck
trans-2-Methyl-2-butenoic acid ≥99%, FG

trans-2-Methyl-2-butenoic acid

trans-2-Methyl-2-butenoic acid ≥99%, FG
Synonym(s):
Tiglic acid, trans-2,3-Dimethylacrylic acid, trans-2-Methyl-2-butenoic acid
Linear Formula:
CH3CH=C(CH3)COOH
CAS Number:
Molecular Weight:
100.12
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing