
Sodium alendronate trihydrate

Synonym(s):
Alendronate sodium trihydrate, 4-amino-1-hydroxy-1-phosphonobutyl phosphonic acid, monosodium, MK-217
Empirical Formula (Hill Notation):
C4H12NaNO7P2 · 3 H2O
CAS Number:
Molecular Weight:
325.12
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing