
Safinamide mesylate salt

Synonym(s):
(S)-(+)-2-[[4-(3-Fluorobenzoxy)benzyl]amino]propanamide; (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propanamide; FCE-26743; PNU-151774E, NW-1015;
Empirical Formula (Hill Notation):
C17H19FN2O2 · xCH4O3S
CAS Number:
Molecular Weight:
302.34 (free base basis)
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing