
Ranolazine Related Compound C

Synonym(s):
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide, N-(2,6-Dimethylphenyl)-1-piperazineacetamide
Empirical Formula (Hill Notation):
C14H21N3O
CAS Number:
Molecular Weight:
247.34
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing