
Proteomix® HIC Propyl-NP5 (5 µm) HPLC Columns

Synonym(s):
Proteomix® HIC Propyl-NP5, Guard Cartridge with Holder
Products
Product Category
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (mm)
Package Size
Particle Size (µm)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing