
Nortriptyline for system suitability

Synonym(s):
Nortriptyline hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-1-propanamine hydrochloride, Desmethylamitriptyline hydrochloride
Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing