
Monoclonal Anti-N-Cadherin antibody produced in mouse

Synonym(s):
Anti-A-CAM, Anti-A-Cell Adhesion Molecule
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing