
Mebrofenin

Synonym(s):
N-{2-[(3-Bromo-2,4,6-trimethylphenyl)amino]-2-oxoethyl}-N-(carboxymethyl)glycine, {{[(3-Bromomesityl)carbamoyl]methyl}imino}diacetic acid
Empirical Formula (Hill Notation):
C15H19BrN2O5
CAS Number:
Molecular Weight:
387.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing