
Flupentixol dihydrochloride

Synonym(s):
4-[3-[2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl]-1-piperazineethanol dihydrochloride
Empirical Formula (Hill Notation):
C23H25F3N2OS · 2HCl
CAS Number:
Molecular Weight:
507.44
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2