
(±)-Methamphetamine-D14 solution

Empirical Formula (Hill Notation):
C10D14HN
CAS Number:
Molecular Weight:
163.32
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique