![Diglycine (glycylglycine) Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/384/301/a3647b16-ef46-4844-8703-b8cbb843fe30/640/a3647b16-ef46-4844-8703-b8cbb843fe30.jpg)
Diglycine (glycylglycine)
![Diglycine (glycylglycine) Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/384/301/a3647b16-ef46-4844-8703-b8cbb843fe30/640/a3647b16-ef46-4844-8703-b8cbb843fe30.jpg)
Synonym(s):
N-Glycylglycine
Empirical Formula (Hill Notation):
C4H8N2O3
CAS Number:
Molecular Weight:
132.12
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing