
Butyl parahydroxybenzoate

Synonym(s):
Butylparaben, 4-Hydroxybenzoic acid butyl ester
Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing