
Bismarck Brown Y

Synonym(s):
4,4′-(m-Phenylenebisazo)bis-m-phenylenediamine dihydrochloride, Basic Brown 1, Bismarck Brown B, Vesuvine
Empirical Formula (Hill Notation):
C18H18N8 · 2HCl
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing