![Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/204/059/39ac29f3-c978-41fd-8537-d0606f1235b1/640/39ac29f3-c978-41fd-8537-d0606f1235b1.png)
Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate
![Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/204/059/39ac29f3-c978-41fd-8537-d0606f1235b1/640/39ac29f3-c978-41fd-8537-d0606f1235b1.png)
Empirical Formula (Hill Notation):
C28H52N2O4
CAS Number:
Molecular Weight:
480.72
Products
Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing