
Anti-SIGLEC6 antibody produced in rabbit

Synonym(s):
Anti-CD327 antigen, Anti-CD33 antigen-like 1, Anti-CDw327, Anti-OB-BP1, Anti-Obesity-binding protein 1, Anti-Sialic acid-binding Ig-like lectin 6 precursor, Anti-Siglec-6
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing