![Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3™ antibody, Mouse monoclonal clone G-A-5, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/189/113/b515fa8e-779d-4871-9810-2754414fac41/640/b515fa8e-779d-4871-9810-2754414fac41.jpg)
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3™ antibody, Mouse monoclonal
![Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3™ antibody, Mouse monoclonal clone G-A-5, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/189/113/b515fa8e-779d-4871-9810-2754414fac41/640/b515fa8e-779d-4871-9810-2754414fac41.jpg)
Synonym(s):
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, Anti-GFAP
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing