Skip to Content
Merck
vibrant-m

Amoxicillin Related Compound D

vibrant-m
Synonym(s):
(4S)-α-[(2R)-2-Amino-2-(4-hydroxyphenyl)acetamido]-4-carboxy-5,5-dimethyl-2-thiazolidineacetic acid monosodium salt, (4S)-2-{[(R)-2-Amino-2-(4-hydroxyphenyl)acetamido](carboxy)methyl}-5,5-dimethylthiazolidine-4-carboxylic acid monosodium salt
Empirical Formula (Hill Notation):
C16H20NaN3O6S
CAS Number:
Molecular Weight:
405.40

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1