Skip to Content
Merck
2-Acetyl-5-methylfuran ≥98%, FG

2-Acetyl-5-methylfuran

2-Acetyl-5-methylfuran ≥98%, FG
Synonym(s):
NSC 80404
Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing