Skip to Content
Merck
(1S,2R)-(+)-Ephedrine-d3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

(1S,2R)-(+)-Ephedrine-d3 hydrochloride solution

(1S,2R)-(+)-Ephedrine-d3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C10H13ClNOD3
CAS Number:
Molecular Weight:
204.71
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique