
1,3,5-Trioxane

Synonym(s):
Metaformaldehyde, Trioxymethylene, sym.-Trioxane, 1,3,5-Trioxane
Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing