Skip to Content
Merck
All Photos(1)

Documents

746274

Sigma-Aldrich

Vinblastine-13C2,d3

99 atom % 13C, 98 atom % D, 97% (CP)

Synonym(s):

13C,d Labeled Vinblastine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C44D3H55N4O9
Molecular Weight:
815.98
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

97% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

2-8°C

SMILES string

O[C@@]1(CC)C[C@H]2C[N@](C1)CCC3=C([C@H](C(OC)=O)C2C4=C(OC)C=C(N(C)C5[C@@]67[C@]8([H])[C@](C=CCN8CC7)(CC)[C@@H](O[13C]([13C]([2H])([2H])[2H])=O)[C@@]5(C(OC)=O)O)C6=C4)NC9=CC=CC=C93

InChI

1S/C46H58N4O9/c1-8-43(54)23-27-24-49(25-43)19-15-29-28-13-10-11-14-32(28)47-37(29)36(38(52)57-6)35(27)30-21-31-33(22-34(30)56-5)48(4)40-45(31)17-20-50-18-12-16-44(9-2,39(45)50)41(59-26(3)51)46(40,55)42(53)58-7/h10-14,16,21-22,27,35-36,39-41,47,54-55H,8-9,15,17-20,23-25H2,1-7H3/t27?,35?,36-,39+,40?,41-,43?,44-,45-,46+/m1/s1/i3+1D3,26+1

InChI key

KPPBSXVQPGVPRR-JDAPZIMESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service