Merck
  • Activated macrophages create lineage-specific microenvironments for pancreatic acinar- and β-cell regeneration in mice.
Activated macrophages create lineage-specific microenvironments for pancreatic acinar- and β-cell regeneration in mice.
Gastroenterology (2014-08-17)
Angela Criscimanna, Gina M Coudriet, George K Gittes, Jon D Piganelli, Farzad Esni
ABSTRACT
Although the cells that contribute to pancreatic regeneration have been widely studied, little is known about the mediators of this process. During tissue regeneration, infiltrating macrophages debride the site of injury and coordinate the repair response. We investigated the role of macrophages in pancreatic regeneration in mice. We used a saporin-conjugated antibody against CD11b to reduce the number of macrophages in mice following diphtheria toxin receptor-mediated cell ablation of pancreatic cells, and evaluated the effects on pancreatic regeneration. We analyzed expression patterns of infiltrating macrophages after cell-specific injury or from the pancreas of nonobese diabetic mice. We developed an in vitro culture system to study the ability of macrophages to induce cell-specific regeneration. Depletion of macrophages impaired pancreatic regeneration. Macrophage polarization, as assessed by expression of tumor necrosis factor-α, interleukin 6, interleukin 10, and CD206, depended on the type of injury. The signals provided by polarized macrophages promoted lineage-specific generation of acinar or endocrine cells. Macrophage from nonobese diabetic mice failed to provide signals necessary for β-cell generation. Macrophages produce cell type-specific signals required for pancreatic regeneration in mice. Additional study of these processes and signals might lead to new approaches for treating type 1 diabetes or pancreatitis.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-α-Amylase antibody produced in rabbit, fractionated antiserum, lyophilized powder