Merck
  • Organocatalytic highly enantioselective synthesis of secondary alpha-hydroxyphosphonates.
Organocatalytic highly enantioselective synthesis of secondary alpha-hydroxyphosphonates.
Organic letters (2006-10-06)
Rajasekhar Dodda, Cong-Gui Zhao
ABSTRACT
[reaction: see text] The first organocatalytic cross aldol reaction of ketones and diethyl formylphosphonate hydrate has been realized by using readily available l-prolinamide as the catalyst. Secondary alpha-hydroxyphosphonates have been synthesized in high enantioselective (up to >99% ee) and good diastereoselectivity.
Sigma-Aldrich
L-Prolinamide, 98%