Merck
  • [Structure of adducts prepared from nicotinamide adenine dinucleotide and their oximes].
[Structure of adducts prepared from nicotinamide adenine dinucleotide and their oximes].
Biochimie (1983-04-01)
J Torreilles, M C Guérin, D Dussossoy, A Crastes de Paulet
ABSTRACT
Preparation of adducts from nicotinamide adenine dinucleotide and a number of oximes is described; these include acetoxime, pyruvatoxime, cyclohexanoxime, cyclopentanoxime. These adducts are closely related to the corresponding NAD-ketone adducts in their spectra properties, but they are stable in acid solutions (pH 5).
Sigma-Aldrich
Acetone oxime, 98%
Sigma-Aldrich
Acetone oxime, purum, ≥98.0% (GC)