Merck
  • Streptococcal intertrigo of the cervical folds in a five-month-old infant.
Streptococcal intertrigo of the cervical folds in a five-month-old infant.
The Pediatric infectious disease journal (2012-05-03)
Robert A Silverman, Richard H Schwartz
ABSTRACT
We present a case of severe intertrigo in the neck of a 5-month-old infant. The cause was Streptococcus pyogenes.
Product Description
Supelco
Cefalexin, VETRANAL®, analytical standard
Cephalexin monohydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cephalexin hydrate, first-generation cephalosporin antibiotic