Merck
  • Comparative study of the effect of baicalin and its natural analogs on neurons with oxygen and glucose deprivation involving innate immune reaction of TLR2/TNFα.
Comparative study of the effect of baicalin and its natural analogs on neurons with oxygen and glucose deprivation involving innate immune reaction of TLR2/TNFα.
Journal of biomedicine & biotechnology (2012-04-27)
Hui-Ying Li, Jun Hu, Shuang Zhao, Zhi-Yi Yuan, Hong-Jiao Wan, Fan Lei, Yi Ding, Dong-Ming Xing, Li-Jun Du
ABSTRACT
This work is to study the baicalin and its three analogs, baicalin, wogonoside, and wogonin, on the protective effect of neuron from oxygen-glucose deprivation (OGD) and toll-like receptor 2 (TLR2) expression in OGD damage. The results showed that baicalin and its three analogs did protect neurons from OGD damage and downregulated protein level of TLR2. D-Glucopyranosiduronic acid on site 7 in the structure played a core of cytotoxicity of these flavonoid analogs. The methoxyl group on carbon 8 of the structure had the relation with TLR2 protein expression, as well as the anti-inflammation. In addition, we detected caspase3 and antioxidation capability, to investigate the effect of four analogs on cell apoptosis and total antioxidation competence in OGD model.
Sigma-Aldrich
Wogonin hydrate, ≥98% (HPLC), solid