Merck
  • Simultaneous determination of 23 amino acids and 7 biogenic amines in fermented food samples by liquid chromatography/quadrupole time-of-flight mass spectrometry.
Simultaneous determination of 23 amino acids and 7 biogenic amines in fermented food samples by liquid chromatography/quadrupole time-of-flight mass spectrometry.
Journal of chromatography. A (2011-11-15)
Shaodong Jia, Yun Pyo Kang, Jeong Hill Park, Jeongmi Lee, Sung Won Kwon
ABSTRACT
A novel liquid chromatography coupled with quadrupole time-of-flight mass spectrometry (LC-Q-TOFMS) method was developed for the simultaneous determination of 23 amino acids and 7 biogenic amines in food samples. These analytes were pre-column derivatized with dansyl chloride and then separated in an Acquity column (1.7 μm; 2.1 mm × 100 mm). The separation of 31 compounds including an internal standard was achieved within 25 min at a flow rate of 0.2 mL/min. The method linearity for each amino acid and biogenic amine had a relatively wide range with r(2)>0.99. The intra- and inter-day precision, expressed as relative standard deviation (RSD), ranged from 1.1 to 4.6% and from 2.0 to 11.2%, respectively. The limit of detection was between 0.005 and 0.4 μg/mL. With a simple dilution, recoveries of around 80-120% were obtained for most of the compounds. No significant matrix effect was observed, and the developed method was successfully applied to the analysis of amino acids and biogenic amines in beer, cheese and sausage samples.
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)